Learn More
ATCC Human herpesvirus 1 DNA; Strain: HF
Supplier:  ATCC VR260D
Encompass
Genomic DNA isolated from a preparation of cell lysate and supernatant from Vero cells (ATCC CCL-81™) infected with Human herpesvirus 1, strain HF (ATCC VR-260™). This product was prepared using methods known to inactivate viruses. It is suitable for use in PCR or other molecular based procedures. Source organism and host cells are also available through the ATCC Catalog.
Catalog No. NC2374164
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.