Learn More
Epredia™ Gemini™ AS Automated Slide Stainer
Improve throughput with the Gemini™ AS Automated Slide Stainer, the ultimate multitasking stainer with a footprint small enough to fit in tight spaces.
Supplier: Epredia™ A81500002CV
Description
The Gemini AS Automated Slide Stainer delivers high-throughput staining capabilities to the busy histology laboratory. With its new intuitive touch-screen interface, operation is easy-to-learn and simple-to-use.
The unique dual-level design, with integrated power supply and touch-screen, ensures maximum space savings. With its small footprint, the Gemini AS Stainer can be placed near the sectioning area for optimal workflow. The intelligent look-ahead control software provides the ability to run up to 12 slide racks simultaneously with minimal reduction in speed. The “urgent” feature enables a rack to be given priority for MOHS surgery or similar STAT needs. The five heated stations provide maximum throughput without the need for a separate drying oven.
Multidimensional Design
- Two-tier design occupies minimal bench space
- Integrated color touch-screen with intuitive software offers quick learning and simple operation
- Scheduler software allows multiple protocols and slide racks to be run simultaneously at optimal speed
- User easily sees progress of slides and placement of reagents with built-in LED lighting
- Fully editable protocols and stain sets provide flexibility in programming
- Fume extraction, battery backup and lockable slide rack ensure safety for the slides and the operator
- 26 or 31 reagent stations accommodate multiple protocols without changing reagents
- One-touch on-screen view of reagent status provides simple reagent monitoring
Performance
- Battery backup with 40 minutes of continual staining power
- Download protocols and log files via USB connectivity
- Load-on-demand flexibility with STAT capability
- Units available with or without five independently heated stations
- Six running water stations
- Four configurable doors for convenient loading and unloading
- Fume control easily managed with carbon filters or vent adapter kit
- Internal flow regulators and anti-siphon devices to protect water supply
- Lockable baskets to ensure secure slides, even upside down
- Auto-return recalls any loaded basket to the load door
- Passcode protection to secure customized protocols
Specifications
Slide Stainer | |
20-slide baskets | |
Stainer: 88 cm; Coverslipper: 50 cm | |
Stainer: 79 cm; Coverslipper: 57.5 cm | |
Stainer: 72 cm; Coverslipper: 64.5 cm | |
50/60 Hz |
Slide Stainer: 79 x 72 x 88 cm (31 x 28 x 35 in.); Coverslipper: 57.5 x 64.5 x 50 cm (22.75 x 25.5 x 19.75 in.) | |
Stainer: 35 in.; Coverslipper: 19.75 in. | |
Stainer: 31 in.; Coverslipper: 22.75 in. | |
Stainer: 28 in.; Coverslipper: 25.5 in. | |
100/240 V | |
Unheated with ClearVue™ Coverslipper (A79200001) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.