Learn More
Hach Company Gelex Secondary Turbidity Standard Kit, 0 - 4000 NTU
Supplier:  Hach Company 2589000
Encompass_Preferred
Gelex Secondary Standards Kit includes 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU, 200-4000 NTU.
Catalog No. 50-199-6275
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.