Learn More
ILEX LIFE SCIENCES LLC GELATIN SOLUTION 0.1 6X500ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0620410X6
Encompass_Preferred
PAN-Biotech Gelatin Solution 0.1% in DPBS, w/o: Ca, Mg (6 x 500 ml). DESCRIPTION: PAN-Biotech Gelatin Solution 0.1% in DPBS, w/o: Ca, Mg for laboratory research applications. Gelatin is a heterogenous mixture of water soluble proteins of high average molecular weight present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, or bones in water. APPLICATION: Applications using gelatin include coating cell culture plates to improve cell attachment for a variety of cell types, addition to PCR to stabilize Taq DNA Polymerase, and use as a blocking agent in Western Blotting, ELISA, and immunohistochemistry. In bacteriology, gelatin can be used as a component of culture media. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-273-3728
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.