Learn More
New England Biolabs, Inc. Gel Loading Dye Purple (6X) – 4 ml
SDP
Supplier:  New England Biolabs, Inc. B7024S
Encompass
Gel Loading Dye, Purple (6X) is the premier gel loading dye from NEB for sharp, tight bands. It is a pre-mixed loading buffer which contains a combination of two dyes, Dye 1 (pink/red) and Dye 2 (blue). The red dye serves as the tracking dye for both agarose and non-denaturing polyacrylamide gel electrophoresis. This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to DNA following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. The dye also contains Ficoll, which creates brighter and tighter bands when compared to glycerol loading dyes. Attention SYBR Safe and GelRed dye users: Due to an increased concentration of SDS in B7024S, NEB recommends using Gel Loading Dye, Purple, No SDS (6X) (NEB# B7025S) instead.
Catalog No. 50-591-186
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.