Learn More
Enzo Life Sciences Gefitinib-based PROTAC 3 (10mg). CAS: 2230821-27-7
Supplier: Enzo Life Sciences ENZCHM2610010
Gefitinib-based PROTAC 3 is targeting on EGFR, consisting of Gefitinib as the warhead linked to VHL-recruiting ligand. Purity: ≥99%. Solubility: Soluble in DMSO (75 mg/mL). Insoluble in water. Long Term Storage: -20°C.