Learn More
Advanced ImmunoChemical, Inc. Disialoganglioside GD2, Human, Lyophilized from Water Solution, 0.5mg
Supplier:  Advanced ImmunoChemical, Inc. 9IG6H0.5MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Neuroscience / Disialoganglioside GD2, human. Source: Human brain tissue. Blood sample from the tissue donors was tested and found negative for HBsAg, HIV-1 and HIV-2 antibodies, HCV, and syphilis. Applications: Prepared from human GD1b ganglioside by enzymatic disintegration of the terminal galactose. Known as a specific cancer ganglioside. The antibodies to the GD2 inhibit cell attachment to various extracellular matrix proteins (collagen, vitronectin, laminin, fibronectin). Analysis: Purity more than 98 % by HPTLC. MW 1649. Presentation: Lyophilized from water solution. Storage Conditions: Before and after reconstitution at -20 °C (between -15 and -30 °C allowed)
Catalog No. 50-153-9826
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.