Learn More
Glass Expansion GazFit Union 6mm (PKT 2)
Supplier: Glass Expansion GAZ06U
GazFit Union 6mm (PKT 2). GazFit Union Connector for 6mm OD laser ablation adaptors and 6mm OD rigid walled tubing. Compatible with laser ablation adaptor 21-809-0965C.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001