Learn More
Cole-Parmer Wika 8736243 GAUGE 2.5IN 100PSI/KPA 1/4 L
Supplier:  Cole-Parmer 6843362
Encompass_Preferred
Our standard pressure gauges are designed for long and reliable service under rugged conditions. Typical applications are for pumps, hydraulic and pneumatic systems, compressors, as a contractor's gauge and for many other applications where the measured media does not corrode brass.
Catalog No. 50-992-225
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.