Learn More
Enzo Life Sciences Ganglioside GT1b . trisodium salt (bovine brain) (5 mg) CAS: 59247-13-1
Supplier: Enzo Life Sciences ALX302011M005
Prevents neurotoxicity of glutamate on neuronal cells. Synonyms: GT1b . 3Na (bovine brain), Trisialoganglioside GT1b . 3Na (bovine brain). Formula: C95H162N5O47 . 3Na. MW: 2126.3 . 69.0 (calculated on sphingosine C18:1 and stearic acid).