Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ G418 Disulfate PhytoPure™
G418 Disulfate PhytoPure™ is an aminoglycoside related to gentamicin that acts as an antibiotic and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths.
$109.00 - $515.00
Specifications
CAS | 108321-42-2 |
---|---|
Color | White |
Form | Powder |
Product Type | Antibiotic |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
02-158-78283
|
MP Biomedicals™
0215878283 |
250 mg |
Each for $109.00
|
|
|||||
02-158-78291
|
MP Biomedicals™
0215878291 |
1 g |
Each for $336.00
|
|
|||||
02-158-78294
|
MP Biomedicals™
0215878294 |
5 g |
Each for $515.00
|
|
|||||
Description
- G418 Disulfate blocks polypeptide synthesis in both prokaryotic and eukaryotic cells and is useful for selection of genetically engineered eukarytoic cells in culture.
- It is an effective selecting agent utilized in killing prokaryotic and eukaryotic cells.
- It is also used for selection and maintenance of prokaryotic and eukaryotic cells transfected with a iNOS promoter construct and neomycin resistance gene.
Specifications
108321-42-2 | |
Powder | |
Non-sterile |
White | |
Antibiotic | |
Cell Culture Grade |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title