Learn More
Chemglass Life Sciences Funnel, Weighing, PP, 2mL Solid Capacity, 1mL Liquid Capacity
Supplier: Chemglass Life Sciences CG176002
Funnel, Weighing, PP, 2mL Solid Capacity, 1mL Liquid Capacity. • Available in anti-static PP, PP and borosilicate glass• With flat bottoms and designed to fit narrow neck vessels