Learn More
Wilmad Labglass FUNNEL ADD CYL GRAD 25ML 14/20
Supplier:  Wilmad Labglass ML1431T702
Encompass_Preferred
Funnel Addition, Graduated, Pressure Equalizing, PTFE Stopcock. Capacity: 25mL; Subdivisions: 0.5mL; Joint Size:14/20; Stopcock Bore: 2. Standard taper Outer joint at top and standard taper joint w/ drip Tube at bottom.
Catalog No. 50-806-695
May include imposed supplier surcharges
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.