Learn More
Enzo Life Sciences FTY-720. HCl (25 mg). CAS: 162359-56-0
SDP
Supplier:  Enzo Life Sciences BMLSL2330025
Encompass_Preferred
Novel immune modulator that alters migration and homing of lymphocytes. Acts as a potent agonist of sphingosine 1-phosphate receptors (S1P) after phosphorylation by sphingosine kinase. Also inhibits cytosolic phospholipase A2 independently of sphingosine 1-phosphate receptors. Alternative name: 2-Amino-2-[2-(4-octylphenyl)ethyl]-1,3-propanediol . HCl, Fingolimod . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml), 100% ethanol or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-201-2145
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.