Learn More
MTC Bio Freezer Box, 5/PK
Supplier:  MTC Bio C2581
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Freezer box, polycarbonate, for 25 (5x5) 5mL tubes
Catalog No. 50-187-1031
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.