Learn More
Electron Microscopy Sciences Formvar/Carbon Film 10 nm/3-4 nm thick on Single 1000 µm hole Nickel Grid
Supplier:  Electron Microscopy Sciences FCFGA1000NISB
Encompass_Preferred
This product was recently added by customer request, and is available for your convenience. We strive to provide our customers with a one-stop shop for the entire scientific supplies category. More relevant content may be added as customer demand increases.
Catalog No. 50-192-6483
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.