Learn More
MilliporeSigma™ Supelco™ Frit for Omnifit™ Column Endpiece
Optimized to avoid clogging and abrasion of chromatographic particles
Supplier: MilliporeSigma™ Supelco™ 56121
Description
- End pieces are equipped with porous frits
- Frits can be made up of sintered metal, glass or plastic
- Can also be used to keep packing material in place
- Easily exchangeable
Specifications
15 mm Bore | |
20 Pk. | |
5 μm |
PTFE | |
Frit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Omnifit is a registered trademark of Diba Industries
Spot an opportunity for improvement?Share a Content Correction