Learn More
Electron Microscopy Sciences EMS Freeze Substitution Kit without Shaker
Supplier: Electron Microscopy Sciences 34500S
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete. With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.