Learn More
Agilent Technologies FP SEAHRSE 96 800U GF 1.2 25CS
Supplier: Agilent Technologies 200929100
Seahorse polypropylene 96-well filter microplate with 1.2um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100