Learn More
Fotodyne™ FOTO/Analyst™ Investigator Workstations
High quality digital photographs of DNA, RNA, and protein gels
Includes:
  • FOTO/Eclipse™ benchtop darkroom
  • CCD camera
  • Transilluminator
  • Image acquisition software
  • Filters
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
F61710PDLNP
View Documents
Fotodyne™
61710PDLNP
Dual light workstation w/8-bit megapixel camera Dual light workstation w/8-bit megapixel camera N/A N/A N/A
F61710PUVNP
View Documents
Fotodyne™
61710PUVNP
UV workstation w/8-bit megapixel camera UV workstation w/8-bit megapixel camera N/A N/A N/A
Description
Description
Versatile systems combine UV or dual-light transilluminator, benchtop darkroom, and CCD camera for seamless gel documentation.
  • Preview images and adjust camera settings while viewing real-time changes on a full-size computer monitor
  • Save images in a variety of file formats, including TIF, BMP, and JPG
  • Flexible integration offers wide range of adjustable exposure settings in 1/30 sec. increments
  • Nanogram-level sensitivity
  • FOTO/Eclipse benchtop darkroom features dual-white light epi-illumination
  • Gliding transilluminator drawer for easy gel positioning, cutting out bands, cleaning
  • Safety-interlocking door prevents accidental UV exposure
  • FOTO/Convertible Dual Light Transilluminator offers 21 × 26cm UV viewing surface covered by hinged electroluminescent white light panel
  • 8-bit, 0.4 megapixel camera is ideal for basic gel documentation, or choose an 8- or 12-bit 1.4 megapixel camera for more demanding applications
Order Info
Electrical Requirements: 120V 60Hz
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.