Learn More
Fotodyne™ FOTO/Analyst™ Investigator/FX Workstations with Manual Zoom, Side Lights, Single Wavelength
Large-format transilluminators provide the flexibility to capture images of many different sample types, including gels, microplates, fluorescent blots, colorimetric blots, TLC plates, and bacterial plates
Includes: 8-48mm (6X) manual zoom lens, manual filter cassette, choice of CCD camera, choice of transilluminator, FOTO/FX darkroom with dual white-light and UV epi-illumination, image acquisition software, PCI frame grabber (with 0.4 megapixel camera) or firewire (with 1.4 and 2.0 megapixel cameras) card, Coomassie™ blue filter (dual-light workstations only), PC, optional thermal printer, all necessary cables and connectors
Products 14
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
16-30D-L
View Documents
Fotodyne™
71630DL
Single-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 12-bit, 1.4 Mpx camera Single-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 12-bit, 1.4 Mpx camera N/A N/A N/A
16-30D-LNP
View Documents
Fotodyne™
71630DLNP
Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 12-bit, 1.4 Mpx camera Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 12-bit, 1.4 Mpx camera N/A N/A N/A
16-30F-DL
View Documents
Fotodyne™
71630FDL
Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx cam Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx cam N/A N/A N/A
16-30F-DLNP
View Documents
Fotodyne™
71630FDLNP
Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera N/A N/A N/A
16-30F-UV
View Documents
Fotodyne™
71630FUV
Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera N/A N/A N/A
16-30F-UVNP
View Documents
Fotodyne™
71630FUVNP
Single-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera Single-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera N/A N/A N/A
16-30U-V
View Documents
Fotodyne™
71630UV
UV transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera UV transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera N/A N/A N/A
16-30U-VNP
View Documents
Fotodyne™
71630UVNP
UV transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera UV transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera N/A N/A N/A
71670DLNP
View Documents
Fotodyne™
71670DLNP
Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 2.0 Mpx camera Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 2.0 Mpx camera N/A N/A N/A
16-70F-DL
View Documents
Fotodyne™
71670FDL
Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx N/A N/A N/A
16-70F-DLNP
View Documents
Fotodyne™
71670FDLNP
Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera N/A N/A N/A
16-70F-UV
View Documents
Fotodyne™
71670FUV
Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera N/A N/A N/A
16-70F-UVNP
View Documents
Fotodyne™
71670FUVNP
Single-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera Single-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera N/A N/A N/A
71670UVNP
View Documents
Fotodyne™
71670UVNP
UV transilluminator; w/PC, w/o printer; 8-bit, 2.0Mpx camera UV transilluminator; w/PC, w/o printer; 8-bit, 2.0Mpx camera N/A N/A N/A
Description
Description
  • Systems are designed to be easily upgraded to include a cooled CCD camera for low-light samples
  • Files can be archived in several formats to preserve data integrity for downstream analysis
  • Workstations can be networked to make files more accessible
  • Manual zoom lens allows adjustments to be made by rotating the on-lens aperature, zoom, and focus rings
  • All systems are PC-compatible
FOTO/FX Benchtop Darkroom
  • Light-tight enclosure houses a CCD camera with zoom lens and filters
  • Flat black interior minimizes stray light
  • Gliding transilluminator platform and side-opening door with interlock provide convenient, safe operation of large-format transilluminators
  • Two interior visible epi-light panels allow easy focusing and even illumination of opaque samples
  • Dual UV epi-lights can be used to optimize sensitivity of samples stained with fluorescent dyes
  • Camera mounting system allows for easy upgrading to other cameras
  • Motorized filter wheel permits filters to be selected through computer software or the darkroom touchpad
FOTO/Flex Transillumators
  • Choose single-wavelength UV; single-wavelength UV, dual-light (UV plus white light); dual-wavelength UV; or dual-wavelength UV, dual-light models
  • UV transilluminators have 25 × 28cm UV glass; four 8 watt, 312nm (peak emission) bulbs; UV transparent protective cover; and preparative and analytical intensity settings
  • White-light transilluminators have a proprietary electroluminescent (EL) visible light panel that offers 15 times more uniform light illumination than traditional bulb-style transilluminators and continuously adjustable intensity control
Image Acquisition Software
  • Integration to 8.3 min. in 1/30 sec. increments
  • Live preview
  • Brightness and contrast
  • Image manipulation: invert, zoom, mirror, rotate, annotate (date, time, text, circle, arrow)
  • Image storage in choice of TIF, JPG, BMP, and PNG formats
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.