Learn More
Fotodyne™ FOTO/Analyst™ Investigator FOTO/Eclipse™ Workstations
Acquire and store electronic images of electrophoresis samples
Supplier:  Fotodyne™ 61530M
Catalog No. FD61530M
Includes: FOTO/Eclipse™ benchtop darkroom; CCD camera with zoom lens, image acquisition software, Investigator controller; thermal printer with one roll of paper; PCI frame grabber/video card; cables, connectors and power cords
Description
Description
Key Features
  • Ideal for obtaining low–light images such as ethidium-bromide–stained DNA gels with as little as 0.1ng per band
  • Capture, manipulate and save images in any of nine file formats, including TIFF, BMP and JPEG
  • Light-tight enclosure features a gliding transilluminator platform and side-opening door with interlock for safe operation
  • Integral dual sidelights allow easy focusing and even illumination of opaque samples such as blots
  • Convenient front-access filter holders enable simple exchange of filters between applications
Image Acquisiton Software
  • All workstations are supplied with real time image acquisition software, live saturation, and on chip integration control
  • Image enhancement tools including background subtraction and sharpen, bright and contrast controls allow the user to invert, zoom, mirror, rotate and annotate (date, time, text, circle, arrow) images
  • Workstations are also available with TotalLab Analysis Software (FD60200) for quantitative image analysis
Dual-Light Transilluminators
  • Dual-Light workstations include FOTO/Convertible™ dual transilluminator and ethidium bromide and Coomassie™ blue interference filters
  • Electrical requirements for all workstations: 120V 60Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.