Learn More
Fotodyne™ FOTO/Analyst™ Investigator Eclipse Dual Light Workstations: 1.4 MPix Camera
Easy-to-use gel doc system quickly captures digital images of gels and saves them as TIF, BMP and JPG formats
Specifications
For Use With (Equipment) | 1D or 2D Complex Gels |
---|---|
For Use With (Application) | 1D or 2D Complex Gels |
Product Type | Dual Light Workstation |
Catalog Number | Mfr. No. | Printer Included | Transilluminator | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Printer Included | Transilluminator | Price | Quantity & Availability | ||||
FD61610DL
|
Fotodyne™
61610DL |
Yes | Single wavelength UV | N/A | N/A | N/A | |||
FD61610DLNP
|
Fotodyne™
61610DLNP |
No | Single wavelength UV | N/A | N/A | N/A | |||
02-580-152
|
Fotodyne™
61610S2DLNP |
No | Dual wavelength UV | N/A | N/A | N/A | |||
02-580-151
|
Fotodyne™
61610S2DL |
Yes | Dual wavelength UV | N/A | N/A | N/A | |||
Description
Offer space-saving FOTO/Eclipse benchtop darkroom plus gliding transilluminator platform for easy access to gels, epi-white light LEDs for framing, focusing and opaque samples, and safety-interlocking door to prevent accidental UV exposure.
- Live-image feature to adjust camera settings while viewing real-time changes on a full-size computer monitor
- Flexible integration feature offers continuously adjustable exposure settings rather than a limited number of presets
- Control to optimize image quality while maximizing sensitivity
- As little as 0.1ng of ethidium bromide-stained DNA can be detected
- Show saturation, crop, label lanes and bands, draw boxes and arrows, and more
- Scientific grade CCD camera with zoom lens and exposure control in 0.1 second increments
- Safety interlock door can be manually overridden to allow band cutting
- Constructed of sturdy 16-gauge steel
Specifications
1D or 2D Complex Gels | |
1D or 2D Complex Gels | |
Dual Light Workstation |