Learn More
Sigma Aldrich Fine Chemicals Biosciences Formaldehyde solution, 50-00-0, MFCD00003274, 100 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences 252549100ML
Linear Formula: HCHO, Molecular Weight: 30.03, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization), Synonym: Formalin. Formaldehyde solution has been used as a fixing agent to fix cells during immunofluorescence imaging and for cross-linking cells during chromatin immunoprecipitation (ChIP) assay. Formaldehyde solution has been used for cross-linking/fixing of cells in ChIP (chromatin immunoprecipitation) assay. It has been used for fixing of cells for imaging.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.