Learn More
Electron Microscopy Sciences Formaldehyde 0.2% Chromic Acid .01% in Sodium Cacodylate Buffer, 7.4 pH 10 ML
Supplier: Electron Microscopy Sciences 15953
Premixed, prebuffered fixatives, are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 ml glass vials (10 ml/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives. Formaldehyde .2% Chromic Acid .1% in Sodium Cacodylate.
pH: 7.4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.