Learn More
Andwin Scientific FORMAGO FORMLIN NEUTRLZER 16CS
Supplier: Andwin Scientific 10852346
The Tissue-Tek FormaGO Formalin Neutralizer features 18% more absorption of formalin and 2-3 times faster neutralization of formalin in as little a 5 minutes. It is sold in a case of 16.