Learn More
Electron Microscopy Sciences Cold Embedding Form Mold, 50 mm
Supplier:  Electron Microscopy Sciences 5101204
Encompass_Preferred
The form mold is a two part mounting cup, designed to be as user-friendly as possible. It has practical handles for removing of the lid and the cylinder is slightly conical for easy removal of the mounts.
Diameter: 50 mm
Catalog No. 50-192-9682
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.