Learn More
Electron Microscopy Sciences Forensic Field Sampler Stub with Adhesive Carbon Tab, clear plastic cover, 8 mm
Supplier:  Electron Microscopy Sciences 7648033
Encompass_Preferred
Glass specimen vial has a high-purity certified 12.7 mm SEM pin stub mounted into the plastic cap. It is ideal for collecting forensic evidence with minimum interference and/or contamination from the sampler. The specimen arrives at the lab ready for carbon coating for SEM/EDX investigation. The Forensic Field Sampler can be securely stored in the glass specimen vial and easily shipped with the cap holding the sampler stub in place. Vial is 25 mm OD x 55 mm high. Adhesive carbon tabs secure the evidence material. Clear plastic cover
Pin length: 8 mm
Catalog No. 50-192-7165
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.