Learn More
Sklar Instruments Econo(TM) Sterile Adson Tissue Forceps, Sklar , 4-3/4 in., 1x2 Teeth, Tissue Forceps, Straight, Smooth Jaws, Stainless Steel, Sterile. CS/50.
SDP
Supplier:  Sklar Instruments 962572
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Econo(TM) Sterile Adson Tissue Forceps, Sklar, 4-3/4 in., 1x2 Teeth, Tissue Forceps, Straight, Smooth Jaws, Stainless Steel, Sterile. Econo(TM) Sterile Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. Econo(TM) Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities. BX50.
Catalog No. NC0958332