Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UF110. 110L - 115 Volt, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert UF110
Encompass_Preferred
Memmert Universal heating and drying Oven - this model is the 115 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 560/480/361-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
Catalog No. 50-225-4285
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.