Learn More
Foamtec CleanWIPE™ Wipers
Open-cell ester-based polyurethane medical-grade foam
Supplier:  Foamtec HT4981
 View more versions of this product
Catalog No. 19-042-474
This item is not returnable. View return policy
Description
Description
Developed for final cleaning of blood oxygenators to prevent harmful fiber contamination.
  • Use as a cushioned work surface for scratch sensitive components
  • Certified non-cytotoxic and lot traceable to comply with FDA standard for the medical device industry
  • Will not interfere with curing of elastomeric compounds, platinum-cured silicones or adhesives
  • Sponge like properties enhance the cleaning and disinfecting of equipment and work surfaces in medical device and pharmaceutical facilities
  • Compatible with gamma, EtO and autoclave sterilization methods
  • Available in small sizes to reduce cost in applications such as catheter wipe down
Specifications
Specifications
30.4 cm
Class 100, FDA
For cleaning of thick screens and substrates, applying or transferring disinfectant, wipe down of medical devices prior to packaging and cleaning or polishing sensitive surfaces, aerospace and medical applications
30.4 cm
CleanWIPE™ cleanroom wipes
625 wipers/case
12 in.
12 x 12 in. (30.4 x 30.4 cm)
12 in.
45PPA Foam
To clean Metal masks and hybrid circuits
0.106 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.