Learn More
Chemglass Life Sciences Funnel, Weighing, 20mL Solid Capacity, 10mL Liquid Capacity, Disposable Antistatic Polypropylene
Supplier: Chemglass Life Sciences CG176008
Funnel, Weighing, 20mL Solid Capacity, 10mL Liquid Capacity, Disposable Antistatic Polypropylene