Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flurofamide, Tocris Bioscience™

Catalog No. 047850
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Urease inhibitor

Flurofamide is a urease inhibitor.

Chemical Identifiers

CAS 70788-28-2
Molecular Formula C7H9FN3O2P
Molecular Weight (g/mol) 217.14
InChI Key QWZFVMCWPLMLTL-UHFFFAOYSA-N
Synonym flurofamide, flurfamide, flurofamida spanish, flurofamine, n-diaminophosphinyl-4-fluorobenzamide, flurofamide usan:inn, flurofamidum inn-latin, flurofamida inn-spanish, unii-599xhu06gh, n-diaminophosphinyl-p-fluorobenzamide
PubChem CID 51173
IUPAC Name N-diaminophosphoryl-4-fluorobenzamide
SMILES C1=CC(=CC=C1C(=O)NP(=O)(N)N)F

Specifications

Quantity 50 mg
Formula Weight 217.14
Percent Purity >98%
Chemical Name or Material Flurofamide
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.