Learn More
STEMCELL Technologies LysoBrite™ Lysosome Dye, Red DND-99, Size: 500 Tests
SDP
Supplier:  STEMCELL Technologies 1000850
Encompass_Preferred
LysoBrite™ dyes are fluorescent dyes that selectively stain lysosomes and other acidic organelles. Due to their hydrophobic nature, LysoBrite™ dyes permeate live cells and selectively accumulate in the lysosomes via the lysosome’s pH gradient. The fluorescence property of these dyes significantly increases within the lysosome’s acidic environment; this fluorescence can be measured using flow cytometry and fluorescence imaging. Due to their selective fluorescence inside lysosomes, LysoBrite™ dyes provide low background and high contrast during imaging. They can be used with proliferating and non-proliferating cells, and are suitable for suspension and adherent cells.
Catalog No. 50-223-1577
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.