Learn More
Enzo Life Sciences Fluorescein-5-maleimide (ultra pure) (25mg). CAS: 75350-46-8
SDP
Supplier:  Enzo Life Sciences ENZ52502
Encompass_Preferred
Fluorescein-5-maleimide enables protein labeling via thiol modifications in most proteins at cysteine residues that either are intrinsically present or resulted from reduction of cystine residues. Unlike iodoacetamides, maleimides do not react with histidine and methionine under physiological conditions. The probe is suitable for assessing conformation- dependent accessibility of cysteine residues in proteins. Wavelength Maxima: Excitation 493nm, Emission 515nm Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2643
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.