Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorene Solution, 5000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1122
Encompass
Click to view available options
Quantity:
1 x 1 mL

Chemical Identifiers

CAS 86-73-7, 67-56-1
Molecular Formula C13H10
Molecular Weight (g/mol) 166.22
MDL Number MFCD00001111
InChI Key NIHNNTQXNPWCJQ-UHFFFAOYSA-N
PubChem CID 6853
ChEBI CHEBI:28266
IUPAC Name 9H-fluorene
SMILES C1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 x 1 mL
CAS Max % 99.36, 0.63
Concentration or Composition (by Analyte or Components) 5000μg/mL
Physical Form Liquid
Chemical Name or Material Fluorene, 5000μg/mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.