Learn More
Cytiva Acrodisc PSF Syringe Filters with PVDF Membrane - 0.2 µm (1000/pkg)
Supplier: Cytiva AP4797
Syringe filters with PVDF membrane are ideal for the filtration of many organic and non-organic solvents in HPLC (0.45 μm) and UHPLC (0.2 μm) sample preparation. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.PVDF syringe filters are chemical compatibility with aggressive acids and alcohols.Acrodisc PSF Syringe Filters have a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of ≤ 150 mL.Available with a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems. The membrane and housing are extremely low in levels of potential extractables and are HPLC certified for low levels of UV-absorbing extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.