Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowtube, Correlated Reading, 150-mm Scale; 49.4 mL/min Air and 0.49 mL/min Water
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0321705
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose 65- or 150-mm flowtubes depending on the selected frame size. Flowtubes are calibrated for direct-reading or correlated reading of air or water at STP (standard temperature and pressure).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.