Learn More
AVANTOR MASTERFLEX (VWR LLC) Hedland DDFXD-A1NA-NN Doppler Flowmeter, 0.15 To 30 FPS, With 4 To 20 mA Input/output
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3261582
Encompass_Preferred
In-field linearization allows adaptation to a wide variety of applications. Accuracy is 2% full-scale over calibrated span. Nonintrusive, clamp-on transducers fit pipes from and above. Clamp-on sensor eliminates the need for in-line flanges, pipe fittings, strainers, or filters.
Catalog No. 50-628-656
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.