Learn More
Electron Microscopy Sciences Cold Embedding Flexible Mold 68 x 37 x 35 mm
Supplier:  Electron Microscopy Sciences 5101255
Encompass_Preferred
This rectangular flexible mold is made of very flexible transparent silicone rubber which makes it very easy to remove the mounts. The inner sides are polished, so the sides of the mounts get clear. The sides are also straight which makes it easier to fasten the mounts in a specimen holder.
Size: 68 x 37 x 35 mm
Catalog No. 50-192-9685
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.