Learn More
Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™
Convenient, pierceable locking lids snap into place with one-handed operation
 View more versions of this product
Catalog No. 02-681-281
Description
Description
Convenient, pierceable locking lids snap into place with one-handed operation
  • Polypropylene tubes with flat tops
  • Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
  • With graduation marks for volume estimation
  • RNase and DNase free
Centrifuges that develop excessive heat may damage this tube.
Specifications
Specifications
RNase, DNase-free
Yes
Assorted
30000xG
Bag
Snap
Polypropylene
0.6mL
Polypropylene
Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
0.1mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.