Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ General Purpose Tweezers, 12cm Long
For precision handling without metal contact.
Description
- Made from polystyrene and high impact polystyrene (67% PS and 33% HIPS)
- Disposable
- Nonsterile
- For handling parts and small samples
- Available with or without tooth
- Available in white and blue colors
Specifications
With tooth | |
Polystyrene and high impact polystyrene (67% PS and 33% HIPS) | |
Tweezers, General Purpose |
Blue | |
120 mm | |
Handling parts and small samples |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction