Learn More
MTC Bio Blu-Lite HS Autoradiography film, 8x10in, 100 sheets/box
Supplier: MTC Bio A8815
This item has a minimum qty of 2 per supplier requirements.
Blu-Lite UHC™ is MTC Bios newest line of high contrast, high sensitivity autoradiography film. Blu-Lite UHCfilm is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications. In addition to Blu-Lite UHCs excellent sensitivity and contrast characteristics, it is also coated on both sides (double emulsion) for improved consistency and overall performance.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.