Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. Supplied stainless steel closure
Supplier: Bellco Glass, Inc. 129038430
Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. Supplied stainless steel closure