Learn More
MilliporeSigma™ Supelco™ EZSafe™ Flash-Chromatography Column with Reservoir and Fritted Disc
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z4159951EA
 View more versions of this product
Catalog No. 11-100-8712
Description
Description
Low pressure (max. 7 psig) medium wall columns with reservoir. Greaseless PTFE insert with GL-45 screw-thread attaches flow control valve to column. Precision all-PTFE valve and drip-tip.
Specifications
Specifications
Flash Chromatography
Flash Chromatography
1000 mL
EZSafe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.