Learn More
New England Biolabs, Inc. Exonuclease I Reaction Buffer – 6 ml
SDP
Supplier:  New England Biolabs, Inc. B0293S
Encompass
This is the NEB reaction buffer supplied with Exonuclease I. New England Biolabs supplies a 10X reaction buffer with all of its enzymes. At a 1X concentration this reaction buffer assures optimal activity of the enzyme.
Catalog No. 50-811-524
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.