Learn More
Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer.
Supplier:  Thermo Scientific™ 10011401
 View more versions of this product
Catalog No. 14-386-420
Explore all special offers
Add to Cart
Description
Description
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer. Combined with a liquid recirculator, this cell changer provides control of sample temperature for up to 7 samples or standards.
  • Easy-to-use sample positioning accessory for use with Thermo Scientific™ Evolution™ 200 series and 300 spectrophotometers.
  • Accessory control fully integrated in Thermo Scientific™ INSIGHT™ and Thermo Scientific™ VISION™ software.
  • Automatic software recognition, smart alignment and serial number reporting
  • Includes carousel and motor base.
  • When placed in both sample and reference beams (Evolution 300 only), this cell changer allows comparison of up to 7 samples with 7 unique references.
  • Monitor the temperature in a cell with Temperature probe accessory for Smart accessories, catalog number 0011401.
  • When deploying this accessory in a regulated or QC environment, a full-system qualification package is available to assure compliance of instrument and accessory as a complete unit.
Specifications
Specifications
Smart Rotary Cell Changer Base
Temperature Probe
Single temperature probe. Connects directly to the Smart accessory motor base. Monitors the temperature in one cuvette.
Temperature probe accessory for Smart accessories
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.