Learn More
Thermo Scientific™ Evolution™ Smart™ Sipper Accessories
Get the most out of your Thermo Scientific™ Evolution™ Spectrophotometer with a wide selection of Thermo Scientific™ Evolution™ Smart™ Sipper Accessories.
Supplier: Thermo Scientific™ 840312000
Description
Thermo Scientific™ Evolution™ Smart™ Sipper Accessories for Thermo Scientific™ Evolution™ Spectrophotometers are easy-to-use sampling accessories featuring a reliable multi-roller peristaltic pump. When the Smart Sipper is installed, all sipper controls are automatically displayed in the software. Methods can be automated so that samples are measured automatically after they are sipped. Smart Sipper Accessories feature modes to return the sample to the original container, send the sample to waste, or run continuously.
- Fully integrated operation with Thermo Scientific™ INSIGHT™ software on all Evolution spectrophotometers. Supported by Thermo Scientific™ VISIONlite™ software on Evolution 200. Samples can be sipped followed by automatic measurement and return of the sample or pumping to waste.
- Automatic recognition by software, smart alignment and serial number reporting.
- Able to handle sample volumes from 200 μL to 9.999 mL and a fully customizable sip volume and air gap.
- Software assisted calibration routine to simplify the process of changing tubing and methods.
- Sippers support 10 mm pathlength cells and shorter pathlength cells in the standard cell holder with use of an appropriate spacer. Swap in a long path cell holder to support flow cells up to 100 mm pathlength.
- Specialized tubing packs available for:
- mild chemical use (silicone tubing)
- acid resistant (c-flex)
- bio-compatible (Chemdurance Bio)
- chemical resistant (Viton)
- Replacement flow-cell connector packs for both Starna and Hellma cells available.
Ordering Alerts:When deploying the Smart Sipper Accessory in a regulated or quality control environment, take advantage of our unique full-system qualification to assure compliance of the instrument and accessory as a complete unit.
Specifications
Works together with Teledyne-Cetac model 280 and 560 autosamplers under INSIGHT Auto control | |
Evolution 350 Spectrophotometers only | |
10 to 100 mm | |
160 μL |
Evolution 350 Smart Sipper Accessory | |
Front panel with sipper beak and manual switch for use with Evolution 350 Spectrophotometer | |
Fully controlled by INSIGHT and INSIGHT Auto software |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.