Learn More
Electron Microscopy Sciences Everlast™ Everlast 247 Rocker
Supplier: Electron Microscopy Sciences 6570009
The Everlast 247 Rocker is made to last. Its powerful motor and heavy-duty drive system are consistent and reliable, even with constant use in cold rooms or incubators. The rocking speed and tilt angle are easily adjustable for gentle or aggressive agitation. It features an extra large 16 x 12" work surface. Platform is made of stainless steel to prevent corrosion or rusting. It will give you years of uninterrupted service.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.