Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 95% Denatured, Vion Biosciences

Catalog No. S02506 Shop All Aldon Products
Click to view available options
Quantity:
1 L
3.8 L
4 x 1 gal.
WARNING: This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
WARNING: This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

This 95% denatured ethanol is a high-quality fixative and dehydrant that allows for optimal staining in histology applications.

  • This 95% denatured ethanol is a high-quality fixative and dehydrant that allows for optimal staining in histology applications.

Chemical Identifiers

CAS 67-63-0, 7732-18-5, 64-17-5, 67-56-1, 108-10-1
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00003568
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Ethanol, Denatured, 95%
PubChem CID 702
ChEBI CHEBI:15235
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Data not available.
Melting Point -114°C
Color Colorless
CAS Min % 8.55%, 0.85%, 80.75%, 3.80%, 5.00%
CAS Max % 81.51%, 4.08%, 0.95%, 5.00%, 8.55%
Appearance Clear, colorless liquid.
Density 0.7919 to 0.7955 g/mL
Boiling Point 74°C to 80°C
Quantity 3.8 L
Explosion Limit 4.00%
Flash Point 5°C
Identification 95% Ethanol
Ignition Point 400°C
Infrared Spectrum Yes
Odor Mild Characteristic Odor
Packaging 1 Gallon Natural HDPE UN Bottle
Physical Form Liquid
Product Line Vion Histology
Assay Percent Range 80.75 to 81.51%
Specific Gravity 0.7919 to 0.7955 g/mL
Linear Formula C2H5OH
Vapor Pressure Ca 50 (at 20°C)
UN Number UN1170
Water 5.00%
Concentration 95%
Formulation 95% Denatured Ethanol
Solubility Information Soluble in water.
InChI Formula 1S/C2H6O/c1-2-3/h3H,2H2,1H3
Percent Purity 80.75 to 81.51%
Grade Histological
Chemical Name or Material Ethanol
Show More Show Less
Hazard Category Toxic if swallowed.
Hazard Statement Toxic if in contact with skin.
Precautionary Statement Causes serious eye irritation.
ShelfLife 3 Years
DOTInformation UN1170, Ethanol, 3(II), Limited quantity equal to or less than 1 L
EINECSNumber 200-578-6
RTECSNumber KQ6300000
Recommended Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.